Borosil® Reagent Square Bottles GL 80 PP Screw Cap PP Pouring Ring, Foxx Life Sciences®
Supplier: Foxx Life Sciences
1506024 1506029 1506033
76490-604CS 1178.08 CAD
76490-604 76490-606 76490-608
Borosil® Reagent Square Bottles GL 80 PP Screw Cap PP Pouring Ring, Foxx Life Sciences®
Bottles Glass Bottles
Borosil® Laboratory bottles are chemically resistant and stable.
  • Mechanically strong and chemical resistant
  • Designed with plastic pouring ring for drip-free operation
  • Screw caps and pouring ring are made of PP
  • Polypropylene pour ring
When fitted with a plastic pouring ring dripping can be totally eliminated. As there is only one size of screw thread for all bottles, the screw caps and pouring rings are fully interchangeable. The bottles, pouring rings and caps are autoclavable and sterilizable.
Certifications: IS 1388 (Part II), EN ISO 4796-2.