VWR® Sterile Masks
Supplier: VWR
VWR®
82011 82012
76470-112CS 1287.62 CAD
76470-112 76470-110
VWR® Sterile Masks
Masks Hygiene Masks
VWR® made in USA sterile masks are an ideal option for single-use needs.
  • Individually bagged
  • Available with earloop or tie configurations
  • 3-ply construction
  • Gamma irradiated to 10⁻⁶
  • Enclosed plastic encapsulated nose wire
A key part of the VWR® made in USA PPE line, these sterile cleanroom masks provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
Packaging: Each mask individually bagged sealed. 10 bags per poly pack. 20 packs per case. Case is poly lined.