Palladium(II) nitrate 2000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Supplier: Ricca Chemical
|
|
RV010134-50N
RCRV010134-50NEA
503.36
CAD
RCRV010134-50N
Palladium(II) nitrate 2000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Palladium (II) nitrate
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Formula:
N₂O₆Pd MW: 230.43 g/mol |
MDL Number:
MFCD00011169 CAS Number: 10102-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...