Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
R1501000-20F R1501000-10F R1501000-1A R1551000-20F R1551000-10F R1601000-20F R1601000-10F R1601000-1A R1501000-4A R1501000-500A R1551000-4A R1551000-500A R1601000-4A 1601-16 R1501000-200PK R1551000-20PK R1551000-1A R1551400-4A R1551400-1A R1601000-200PK
RC1501-5EA 630.42 CAD
RC1501-5 RC150125 RC150132 RC1551-5 RC155125 RC1601-5 RC160125 RC160132 RC1501-1 RC1501-16 RC1551-1 RC1551-16 RC1601-1 RC1601-16 RC1501-20B RC1551-20 RC1551-32 RC1551.4-1 RC1551.4-32 RC1601-20B
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Buffers pH Calibration Buffers
  • Certified traceable to NIST Standard Reference Materials
  • Lot numbers and expiration dates