LiquidGuard™ Sterile DuraGrip Knee-High Boot Covers, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
SGR2795WHS/M-E SGR2795WHL/XL-E SGR2795WH2XL-E
76488-328CS 613.42 CAD
76488-328 76488-330 76488-332
LiquidGuard™ Sterile DuraGrip Knee-High Boot Covers, Apex Aseptic Products
Disposable Garments
Sterile DuraGrip LiquidGuard™ knee-high boot covers are suitable for cleanroom use.
  • Impervious yet breathable
  • Low particle shedding, rated category 1 per Helmke Drum Test
  • Latex-free, TSE-free, BSE-free
  • Manufactured in a controlled environment
  • Gamma irradiated validated sterile to SAL 10⁻⁶
  • Suitable to use in ISO Class 5 or higher cleanrooms
Boot cover soles are made with specially engineered DuraGrip PVC material, which is ultra-non-skid and durable. The upper part of the boot cover is made with LiquidGuard™ fabrics. Tunneled elastic sewing around the ankle and knee reduces particle shedding. The garments are specially folded to ensure proper aseptic donning.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected.
Pass Blood Penetration Test per ASTM F1670.
Delivery information: Each package includes a gamma irradiation indicator dot, and each shipping case is affixed with a Certificate of Conformance and Certificate of Sterility.
Packaging: Boot covers are packaged 2 pieces per package, triple bagged, 200 pieces per case.