Sterile Full Face Shield, Apex Aseptic Products
Supplier: Apex
S5850F-SB
76488-348CS 496.04 CAD
76488-348
Sterile Full Face Shield, Apex Aseptic Products
Face Shields
Optically clear and anti-fog treated plastic shields provide full face protection against fluid and splashes onto the face and eyes.
  • Comfortable forehead foam cushion prevents fluid splashes from above
  • Low particle shedding, rated category 1 per Helmke Drum Test
  • Latex-free, TSE-free, BSE-free
  • Manufactured in a controlled environment
  • Gamma irradiated validated sterile to SAL 10⁻⁶
  • Suitable to use in ISO Class 5 or higher cleanrooms
Each face shield is individually packaged with a gamma irradiation indicator dot.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected.
Delivery information: Each case includes a Certificate of Conformance and Certificate of Sterility.
Packaging: Face shields are packaged 1 piece per package, triple bagged, 100 pieces per case.