LiquidGuard™ Plus Sterile Cleanroom Coveralls with Attached Hood and Safe-Grip Booties, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
SGR4903PLAWHS SGR4903PLAWHM SGR4903PLAWHL SGR4903PLAWHXL SGR4903PLAWH2XL SGR4903PLAWH3XL SGR4903PLAWH4XL
76488-284CS 544.37 CAD
76488-284 76488-286 76488-288 76488-290 76488-292 76488-294 76488-296
LiquidGuard™ Plus Sterile Cleanroom Coveralls with Attached Hood and Safe-Grip Booties, Apex Aseptic Products
Disposable Garments
Sterile LiquidGuard™ Plus cleanroom coveralls feature a zipper front closure, with an attached hood and attached Safe-Grip booties.
  • Made with high strength LiquidGuard™ Plus microporous fabrics
  • Pass Blood Penetration Test per ASTM F1670
  • Pass ChemoDrug Permeation Test per ASTM F739-12
  • Anti-static treated, latex-free, TSE-free, and BSE-free
  • Easy breathability and comfortable to wear
  • Zipper front, coveralls attached with crown hood to minimize open face exposure
  • Coverall attached with Ultra-Non-Skid SafeGrip booties to prevent accidental falls
  • Low particle shedding, rated category 1 per Helmke Drum Test
  • Gamma Irradiated Validated Sterile to SAL 10⁻⁶
  • Suitable to use in ISO Class 5 or higher cleanrooms
Inside seams are bonded with Teal color fabrics to indicate areas safe to touch. Tunneled elastic cuffs with thumb loops help keep sleeves in place and prevent bare arms from exposure. Uniquely designed Clean-Don™ gowning system incorporates inside out folding and telescope leg method to ensure wearer’s hands are not contaminating the outside of the sterile garments and coverall’s legs are not touching the floor.
Each piece is individually packaged, triple bagged in plastic bags with easy tear notches, and labeled with item code, product name, lot number, production date, expiration date, and fixed with gamma irradiation indicator dot. Manufactured in a controlled environment.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 Certified and FDA registered and inspected. Pass Blood Penetration Test per ASTM F1670. Pass ChomoDrug Permeation Test per ASTM F739-12.
Delivery information: Each case includes a Certificate of Conformance and Certificate of Sterility.
Packaging: Coverealls are packaged 1 piece per package, triple bagged, 25 pieces per case.