Rotational Viscometers, ROTAVISC HELI Complete Series, IKA
Supplier: IKA
25008214 25008218 25008216 25008217
76499-014EA 7210.96 CAD
76499-014 76499-012 76499-018 76499-016
Rotational Viscometers, ROTAVISC HELI Complete Series, IKA
Viscometers Rotational Viscometers
The ROTAVISC HELI series determines the viscosity of liquids in all areas of application ranging from the laboratory to quality control. The four devices measure in different viscosity ranges.
  • Large TFT display allows for an intuitive menu guidance
  • Digital spirit level supports the correct set-up of the start-up condition
  • Ramp function simplifies repetitive tasks
  • Very high measurement accuracy
  • Stepless speed adjustment
  • Timer and graph function
  • HELISTAND moves the mounted viscometer and its rotating body slowly and uniformly up and down in the sample
The level of measurement accuracy of the ROTAVISC HELI rotational viscometer for both Newtonian and non-Newtonian fluids is ±1% of the measuring range. The reproducibility is ±0,2%.
Ordering information: ROTAVISC HELI complete packages include SP 1 - 4 (lo-vi)/SP 7 - 12 (me-vi, hi-vi I, hi-vi II), protective bracket, temperature sensor, quick connector, hook connector and HELISTAND stand including a T-spindle set.