Sera- Separa Columns, Caplugs
Supplier: Caplugs
208-3084-020 208-3193-020
76498-808CT 970.18 CAD
76498-808 76498-810
Sera- Separa Columns, Caplugs
Chromatography Columns
Sera-Separa columns are ideal for use in removing fibrin and macro particles from serum to prevent line clogging.
  • Graduated barrel tells exactly how much liquid has been filtered
  • Wide-body style prevents sample probes from hitting side walls
Users insert the 16 mm flange into the evacuated blood tube and push downward.