Surety Columns, Caplugs
Supplier: Caplugs
208-3042-020 208-3045-020
76498-800CT 273.86 CAD
76498-800 76498-802
Surety Columns, Caplugs
Chromatography Columns
Surety columns are ideal for use in removing fibrin and macro particles from serum to prevent line clogging.
  • Surety columns have integral caps that fit directly onto sample cups
Choose from sizes ranging in size from 2.4 to 5.0 ml capacity.
Order Now
SPECIFICATIONS
Environmentally Preferable