ESD Safe Microscope Covers, Transforming Technologies
Supplier: Transforming Technologies
MC1221W
76474-668EA 61.6 CAD
76474-668
ESD Safe Microscope Covers, Transforming Technologies
Surface Protectors Equipment Protectors
Protect microscope optics from dust and other contamination while keeping the work station ESD safe.
  • Permanently anti-static: less than 10⁶
  • Made with polyester and conductive yarn
  • Fits most common microscopes
Common microscope covers are made from vinyl, a material that is an insulator and potential static generator. This cover is made from ESD safe materials and does not generate a significant charge.