Warmbier SRM200 Pocket Digital Surface Resistance Verification Kit, Transforming Technologies
Supplier: Transforming Technologies
7100.SRM200.VK
76474-630EA 5235.27 CAD
76474-630
Warmbier SRM200 Pocket Digital Surface Resistance Verification Kit, Transforming Technologies
Resistance Testers
Full-featured digital Surface Resistance Meter with lightweight travel probes, two-point probe, data storage, temperature, and humidity testing.
  • Perform resistance measurements to determine if a surface is dissipative, conductive, or insulative
  • Test range: 10³ to 10¹²
  • Store test data that can be accessed via USB
  • Measures temperature and humidity
  • Test voltage: 10 /100 V (automatic ranging)
  • Built-in resistivity probes
  • Carrying case and calibration certificate included
SRM200 is a very useful instrument for checking surface resistance and resistance to ground. The instrument can be used on different work surfaces, floors, chairs, carts, clothing, and packaging materials, etc. With the additional 'two-point probe' adapter also Rpp measurements can be done on small areas like in cavities of vaccum formed trays. The measured value is displayed via LED that indicates whether the value is conductive, dissipative, or insulating. All readings can be stored in the instrument and can be easily transferred to PC via USB cable using the included software.