Warmbier EFM51 Electrostatic Field Meters, Transforming Technologies
Supplier: Transforming Technologies
7100.EFM51 7100.EFM51.CPS EFM51.CPS.SET EFM51.PLUS 7100.EFM51.VK EFM51.WT
76474-616EA 4025.62 CAD
76474-616 76474-618 76474-620 76474-622 76474-624 76474-626
Warmbier EFM51 Electrostatic Field Meters, Transforming Technologies
Electrical Meters Voltmeters
A convenient hand-held device designed to locate and measure electrostatic fields on surfaces.
  • Measures static voltages within ±20 kV (20000 V)
  • Two LED guide ring lights help position the field meter
  • Storage bag including ground cable and clip
  • Pocket-size, portable design
  • Battery operated
The EFM51 is a full-featured, hand-held field meter used to perform static surveys and evaluate charge accumulation in electronic manufacturing and industrial operations. The EFM51 measures static voltages within ±20 kV (20000 V) with a microcontroller thatMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable