J.T.Baker® Endonuclease

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

BAKR7108-05 BAKR7108-10 7108-40 7108-50
CAJT7108-01EA 158 CAD
CAJT7108-01 CAJT7108-05 CAJT7108-10 JT7108-40 JT7108-50
J.T.Baker® Endonuclease
Enzymes

J.T.Baker® Endonuclease Ultrapure Bioreagent is designed for the degradation of both single stranded and double stranded DNA and RNA. J.T.Baker® Endonuclease Ultrapure Bioreagent is used to ensure host cell DNA impurities are removed; driving process efficiency by lowering viscosity and preventing aggregation. J.T.Baker® Endonuclease is an enzyme based upon the native endonuclease of Serratia marcescens, enabling rapid clearance of residual DNA and RNA during the production and purification of both recombinant proteins and viral vectors. Non-animal origin. The purity of materials in non-negotiable. J.T.Baker® Endonuclease Ultrapure Bioreagent acts to degrade and eliminate extraneous genetic material, ensuring the pristine quality of your final product.


  • High purity, bioburden and endotoxin tested
  • Rapid clearance of residual DNA and RNA
  • Easily cleared in downstream processing, detectable by routine analytical methods
  • 0.2 µm filtered solution
  • Various size offerings
  • Proven performance with J.T.Baker® Workflow solutions


Appearance: Passes Test
Identification: Passes Test
Purity by SDS-PAGE: ≥ 90%
Activity: ≥ 250 U/µL
Specific Activity: ≥ 1.1 x 10^6 U/mg
Total Aerobic Microbial Count: ≤ 10 cfu/ 100,000U
Total Yeast and Mold Count: ≤ 10 cfu/ 100,000U
Endotoxin: For Information Only

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR