Viral inactivation solution, Ultrapure Bioreagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
J.T.Baker® Viral Inactivation Solution Ultrapure bioreagent is a highly effective, ready-to-use product, specifically developed for detergent-based viral inactivation in biopharmaceutical operations and plasma derived therapy processes. J.T.Baker® Viral Inactivation Solution is formulated with non-ionic detergent, that is readily biodegradable and alkylphenol ethoxylate free, addressing environmental concerns associated with REACH. This innovative solution is optimized to have low protein reactive impurities, low viscosity and compatibility with typical downstream processes. J.T.Baker® Viral Inactivation Solution Ultrapure bioreagent is available in a variety of sizes to support development to large scale commercial batches. Detail description provided in technical literature.
- High purity, tested for bioburden and impurity
- Effective inactivation of virus
- Easily cleared in downstream processing, detectable to 1ppm
- Readily biodegradable
- 0.2 µm filtered solution
Proprietary Formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)
Specification Test Results
Identification (by FTIR) | Passes Test |
Appearance | Passes Test |
Purity by HPLC | min 95% |
Assay | Passes Test |
Acid value | max 10 |
Viscosity (25°C) | For information only |
Density | 0.9 – 1.1 g/mL |
Limit of free ethylene oxide (Target <1 ppm) | For information only |
Total microbial count (No growth detected) | Passes Test |
Endotoxin | For information only |
Protein reactive impurities (100x Diluted) | For information only |
Arsenic (As) | For information only |
Cadmium (Cd) | For information only |
Lead (Pb) | For information only |
Mercury (Hg) | For information only |
Cobalt (Co) | For information only |
Nickel (Ni) | For information only |
Vanadium (V) | For information only |
Silver (Ag) | For information only |
Gold (Au) | For information only |
Iridium (Ir) | For information only |
Osmium (Os) | For information only |
Palladium (Pd) | For information only |
Platinum (Pt) | For information only |
Rhodium (Rh) | For information only |
Ruthenium (Ru) | For information only |
Selenium (Se) | For information only |
Thallium (Tl) | For information only |
Barium (Ba) | For information only |
Chromium (Cr) | For information only |
Copper (Cu) | For information only |
Lithium (Li) | For information only |
Molybdenum (Mo) | For information only |
Antimony (Sb) | For information only |
Tin (Sn) | For information only |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...