Anti-HSV2 Mouse Balb/c Monoclonal Antibody [clone: DBM15.69]
Supplier: Rockland Immunochemical
MOB542-01 MOB542-01
76539-418EA 893.05 CAD
76539-418 CA76539-418
Anti-HSV2 Mouse Balb/c Monoclonal Antibody [clone: DBM15.69]
Antibodies
Anti-Herpes Simplex Virus Type II (HSV2) (MOUSE) Monoclonal Antibody was tested with IHC-P positive control infected lung tissues. Cellular Localization: Cytoplasmic, nuclear. The user is advised to validate the use of the products with their tissue specimens prepared and handled in accordance with their laboratory practices. Consult references (Kiernan, 1981: Sheehan & Hrapchak, 1980) for further details on specimen preparation.
  • Reacts with herpes simplex virus type 2
  • Specific for the viral glycoprotein D (gD) protein
  • Tested with IHC-P positive
More Product Information