Elastic Fabric Adjustable Wrist Strap Sets, WB8000 Series, Transforming Technologies
Supplier: Transforming Technologies
WB8016 WB8037 WB8043
76474-824EA 9.51 CAD
76474-824 76474-826 76474-828
Elastic Fabric Adjustable Wrist Strap Sets, WB8000 Series, Transforming Technologies
Grounders Wrist Grounders
ESD wrist strap set is a comfortable ESD grounding device for industrial and controlled environments.
  • Unique locking clasp eliminates excess band material when tightened
  • Soft woven fabric of the wrist band allows the skin to breathe
  • Highly conductive copper sulfide thread for conductivity
  • Band expands to fit most wrists
  • 4 mm snap fits most common coil cords
The WB8000 Series Wrist Band and Coil Cord set features an elastic adjustable band with a unique locking clasp that eliminates excess wrist band material. The band is standard with 4 mm, machined male snap and provides a 360° conductive contact around the wrist, providing maximum skin contact. The Band Set is completed with a premium coil cord featuring 2.5 mm polyurethane coil insulation with excellent coil memory. A shorter, swivel type banana jack increases coil life, prevents cord tangles, and fits most bench grounds. It is uses highly conductive copper sulfide nylon thread for conductivity and it has a maximum resistance of 160 ohms.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and ESDA Standard 1.1-2006.