Dual Conductor Fabric Wrist Band and Coil Cord Sets, WB7000 Series, Transforming Technologies
Supplier: Transforming Technologies
WB7050 WB7050R WB7100 WB7100R WB7200 WB7200R
76474-812EA 66.45 CAD
76474-812 76474-814 76474-816 76474-818 76474-820 76474-822
Dual Conductor Fabric Wrist Band and Coil Cord Sets, WB7000 Series, Transforming Technologies
Grounders Wrist Grounders
The one-size-fits-all Dual Conductor Fabric Wrist Bands are designed for use with Dual Conductor Constant Work Station monitors.
  • Dual-Wire conductor provides redundancy
  • Superior contact with the skin for reliable grounding
  • Available with standard and right angle cords
The wrist strap features a silver plated, monofilament, continuous thread woven together with elastic nylon for full conductivity and comfort. A double insulated jacket provides incredible durability and unmatched reliability via a redundant grounding path.
Certifications: MeetsMore Product Information