Dual Conductor Metal Wrist Strap Sets, WB5000 Series, Transforming Technologies
Supplier: Transforming Technologies
WB5100LR WB5050S WB5200MR WB5100M WB5050M WB5050L WB5200LR WB5200S WB5050LR WB5200M WB5050MR WB5200L WB5200SR WB5100SR WB5100MR WB5100S WB5050SR WB5100L
76474-790EA 135.53 CAD
76474-790 76474-794 76474-772 76474-782 76474-770 76474-792 76474-780 76474-776 76474-764 76474-786 76474-796 76474-774 76474-762 76474-784 76474-768 76474-778 76474-766 76474-788
Dual Conductor Metal Wrist Strap Sets, WB5000 Series, Transforming Technologies
Grounders Wrist Grounders
The adjustable WB5000 series dual conductor wrist strap set is designed for use with a Dual Conductor Constant Workstation Monitor to maximize ESD protection.
  • Premium metal expansion wrist strap with more reliable path-to-ground
  • Superior contact with skin
  • Dual-Wire conductor provides redundancy
  • Fixed size bands available in three sizes
Adjustable Dual Wire Wrist Strap sets provide redundancy by including two connections to ground, so even if one dual wire wrist strap conductor is severed, the operator has reliable path-to-ground with other wire. The Speidel metal band features two 180° conductive paths around the wrist, providing maximum skin contact. The expandable band is constructed with two stainless steel back plates and is available in small, medium, and large sizes. A double insulated jacket provides incredible durability and unmatched reliability.
Certifications: Meets requirements of ANSI/ESD S20.20 and ANSI/ESD S1.1.