Anti-Allergy Wrist Band and Coil Cord Sets, WB5600 Series, Transforming Technologies
Supplier: Transforming Technologies
WB5026 WB5637 WB5643 WB569020
76474-760EA 8.97 CAD
76474-760 76474-800 76474-802 76474-804
Anti-Allergy Wrist Band and Coil Cord Sets, WB5600 Series, Transforming Technologies
Grounders Wrist Grounders
This Anti-allergy ESD wrist strap set is the ideal grounding device for people with allergies to common metals.
  • No metal contacts the skin, unlike common wrist straps that use a metal plate
  • Hypoallergenic silver-plated nylon thread for conductivity
  • Back plate has a plastic cover so that metal does not meet skin
Anti-allergy wrist band sets feature hypoallergenic silver-plated nylon thread for conductivity and a plastic covered backplate so that metal does not meet skin. The wrist band and coil cord set feature a 360° conductive contact around theMore Product Information