BenchMasher 400 Paddle Blender, Benchmark Scientific
Supplier: Benchmark Scientific
Benchmark®
IPD4400 IPD4400-E
76498-744EA 8731.59 CAD
76498-744 76498-742
BenchMasher 400 Paddle Blender, Benchmark Scientific
Homogenizers Laboratory Paddle Blenders
Adjustable paddles, chamber temperature control and UV decontamination make the BenchMasher 400 ideal for homogenizing laboratory samples.
  • Contamination free mixing/homogenization of samples
  • Touch screen control
  • Adjustable stroke length
  • Built in UV light for chamber decontamination
The BenchMasher provides fast and effective mixing, ensuring consistent homogenization. It is ideal for preparation of samples were eliminating contamination is critical, such as pathogen and microbial detection in food and water filters. However, it’s use is not limited to those areas – the BenchMasher has applications anywhere larger solid samples need broken down or products such as powders and polymers need mixed.
Samples up to 400 ml are conveniently loaded in the sterile blender bags which are sealed upon closing the door. Mixing paddles rapidly strike the bags, producing thorough mixing and homogenization without ever directly contacting the sample. Programming is user friendly - mixing speed and cycle time are easily adjusted on the large, full color touch screen. Paddle position can be adjusted to ensure efficient processing of samples. For applications requiring thermal regulation (such as Trichinella testing in meat) the chamber can be heated and maintained at temperatures up to 65 °C.
The door is removable for access to the chamber and stainless steel mixing paddles. Each paddle includes a convenient rubber cover that is easily removed or replaced. A UV light (254 nm) is built into the chamber and can be activated at any time to perform complete decontamination.
Ordering information: Bags sold separately.