Dual Conductor Metal Wrist Band and Coil Cord Sets with Parking Jack, WB2695P Series, Transforming Technologies
Supplier: Transforming Technologies
WB2695P10 WB2695P20 WB2695P5
76474-744EA 64.02 CAD
76474-744 76474-746 76474-748
Dual Conductor Metal Wrist Band and Coil Cord Sets with Parking Jack, WB2695P Series, Transforming Technologies
Grounders Wrist Grounders
The adjustable WB2695P series dual wire fabric wrist strap set features two 180° conductive paths around the wrist, providing maximum skin contact and two 4 mm male snaps.
  • Adjustable fabric wrist strap with 5, 10, or 20' coil cord
  • 'One Snap' design with two 4mm snaps enclosed in a molded head
  • Equipped with convenient parking jack for the cord when not in use
  • Dual-Wire conductor provides redundancy
Constructed with stainless steel back plates, nylon molding, and a premium elastic yarn that provides excellent fit for comfort and long life. The series polyurethane coil insulation and strain relief molding offers excellent coil memory and endurance. The molded snap head provides a convenient parking jack for the cord when not in use.
Certifications: Meets requirements of ANSI/ESD S20.20 and ANSI/ESD S1.1.