VinylStat D Homogeneous Vinyl ESD Table Mats, Transforming Technologies
Supplier: Transforming Technologies
VMD2450B VMD2450GY VMD3050B VMD3050GY VMD3650B VMD3650GY VMD4850B VMD4850GY
76474-710EA 1346.11 CAD
76474-710 76474-712 76474-714 76474-716 76474-718 76474-720 76474-722 76474-724
VinylStat D Homogeneous Vinyl ESD Table Mats, Transforming Technologies
Surface Protectors Bench Protectors
Single-layer vinyl homogeneous ESD work surface mat is ideal for general use.
  • Surface resistivity is in the dissipative range: 10⁷ to 10⁸
  • Homogeneous polymer mix construction
  • Available in blue or grey; supplied as mat rolls
A homogeneous polymer mix construction with unsurpassed electrical properties, gives VinylStat D a consistent, controlled static drain and physical properties offering the highest level of protection. VinylStat D is made with the same homogeneous polymer mix as the VinylStat C mats but are 0.0625" thick.
Resistant to degradationMore Product Information