VinylStat C Homogeneous Vinyl ESD Table Mats, Transforming Technologies
Supplier: Transforming Technologies
VMC2450B VMC2450GY VMC3050B VMC3050GY VMC3650B VMC3650GY VMC4850B VMC4850GY
76474-694EA 1835.57 CAD
76474-694 76474-696 76474-698 76474-700 76474-702 76474-704 76474-706 76474-708
VinylStat C Homogeneous Vinyl ESD Table Mats, Transforming Technologies
Surface Protectors Bench Protectors
Single-layer vinyl homogeneous ESD work surface mat is ideal for general use.
  • Surface resistivity is in the dissipative range: 10⁷ to 10⁸
  • Homogeneous polymer mix construction
  • Available in blue or grey; supplied as mat rolls
A homogeneous polymer mix construction with unsurpassed electrical properties gives VinylStat C a consistent, controlled static drain and physical properties offering the highest level of protection. Available in blue or grey.
Resistant to degradation by inorganic acids, organic acids, reducing agents, detergent solutions, alcohols, aliphatic hydrocarbons, mineral oil, amines, and aldehydes.