VinylStat B Three-Layer ESD Table Mat with Foam Back, Transforming Technologies
Supplier: Transforming Technologies
VMB2450B VMB2450GY VMB3050B VMB3050GY VMB3650B VMB3650GY VMB4850B VMB4850GY
76474-678EA 2080.37 CAD
76474-678 76474-680 76474-682 76474-684 76474-686 76474-688 76474-690 76474-692
VinylStat B Three-Layer ESD Table Mat with Foam Back, Transforming Technologies
Surface Protectors Bench Protectors
VinylStat B provides excellent ESD protection and extra cushion for component safety and operator comfort.
  • Provides extra cushion for component safety and operator comfort
  • Three layers of vinyl with excellent ESD protection
  • Available in blue or grey; supplied as mat rolls
Table mats utilize a three-layer construction consisting of a conductive metalized fleece sandwiched between a solid, vinyl top surface and a foam, base layer. Surface resistivity is in the dissipative range: 10⁷ to 10⁸.
Certifications: Meets or exceeds requirements of ANSI/ESD S20.20 per ANSI/ESD STM4.1.