HyFlex® 11-755 Light-Duty Industrial Gloves, Ansell
Supplier: Ansell Canada
11755060 11755070 11755080 11755090 11755100 11755110 11755120
CA76478-892CS 3910.71 CAD
CA76478-892 CA76478-894 CA76478-896 CA76478-898 CA76478-900 CA76478-902 CA76478-904
HyFlex® 11-755 Light-Duty Industrial Gloves, Ansell
Gloves
Ultra-thin ANSI Cut level A5 / ISO Cut level E gloves with 4× more cut resistance for unmatched protection.
  • Ultra-thin breathable gloves offer high cut resistance, while maintaining enhanced flexibility and tactility
  • Touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks
  • Designed to fit the natural shape of the hand to help reduce hand fatigue
  • Clean and skin friendly
  • Dermatologically approved by Dermatest®
Lightest weight ANSI A4/ ISO Cut D glove featuring INTERCEPT™ Technology, the ultra-lightweight design provides dexterity for handling and assembly of sharp parts. The 11-755 series is recommended for applications including component inspection and assembly, picking and fastening, final assembly, and applying finish in automotive, machinery, and metal fabrication industries.
Certifications: REACH compliant. EN 388 3X41E. ANSI 4 ABR. ANSI A5 CUT.