HyFlex® 11-754 Light-Duty Industrial Gloves, Ansell
Supplier: Ansell Canada
11754060 11754070 11754080 11754090 11754100 11754110 11754120
CA76478-878CS 3647.65 CAD
CA76478-878 CA76478-880 CA76478-882 CA76478-884 CA76478-886 CA76478-888 CA76478-890
HyFlex® 11-754 Light-Duty Industrial Gloves, Ansell
Gloves
Lightest weight ANSI A4/ ISO Cut D glove featuring INTERCEPT™ Technology, these ultra-thin breathable gloves offer high cut resistance, while maintaining enhanced flexibility and tactility.
  • Ultra-thin ANSI Cut level A4 / ISO Cut level D gloves
  • 3x more cut resistance for unmatched protection
  • Touchscreen compatible for enhanced productivity
  • Ultra-lightweight design provides dexterity for handling and assembly of sharp parts
Ultra-thin breathable gloves are touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks. Designed to fit the natural shape of the hand to help reduce hand fatigue. Clean and skin friendly, HyFlex® 11-754 gloves are dermatologically approved by Dermatest®.
Certifications: REACH compliant. EN 388 3X41D. ANSI 4 ABR. ANSI A4 CUT.