Control Expansion
Supplier: Oxford Nanopore Technologies
EXP-CTL001 EXP-RCS001
76487-110EA 118.5 CAD
76487-110 76848-052
Control Expansion
Nucleic Acid Reagents Next Generation Sequencing Reagents
DNA or RNA for sequencing control experiments.
  • Perform DNA or RNA control experiments to familiarise yourself with nanopore sequencing
DNA Control Expansion (EXP-CTL001): Lambda DNA is 48 kb long and serves as a good model system to evaluate the nanopore sequencing workflow. Control experiments should be carried out when users are getting started with nanopore sequencing devices, or when troubleshooting library preparation. Pack of six vials. Contains Lambda DNA (LMD), sufficient for at least 24 reactions.
RNA Control Expansion (EXP-RCS001): contains RNA control sample allowing control runs to support Oxford Nanopore cDNA sequencing kits. Pack of three vials. Contains sufficient control RNA for six reactions.
Delivery information: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.