PCR Expansion, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
EXP-PCA001
76487-126EA 758.4 CAD
76487-126
PCR Expansion, Oxford Nanopore Technologies
Nucleic Acid Reagents Next Generation Sequencing Reagents
PCR Primers (PRM) and PCR Adapters (PCA).
  • Additional PCR primers and adapters for use with Ligation Sequencing Kits
This kit contains PCR Primers and PCR Adapters, sufficient for at least 24 reactions, and is compatible for use with the Ligation Sequencing Kits (76487-140 and 76487-138).
Delivery information: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.