Direct RNA Sequencing Kit, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
SQK-RNA002 SQK-RNA004
76487-164EA 1012.78 CAD
76487-164 77308-582
Direct RNA Sequencing Kit, Oxford Nanopore Technologies
Nucleic Acid Reagents Next Generation Sequencing Reagents
A sequencing kit optimised for sequencing native RNA with improved output and accuracy.
  • Sequence native RNA not a copy — eliminate PCR bias
  • Characterise challenging transcripts that are difficult to reverse transcribe
  • Directly explore the attributes of native RNA such as modified bases
The Direct RNA Sequencing Kit is used to prepare and sequence native RNA without conversion to cDNA. Inputs include poly(A)-tailed RNA or total RNA, such as eukaryotic mRNA and viral RNA.
The latest Direct RNA Sequencing Kit upgrade (SQK-RNA004) includes increased sequencing output and improved modal raw read accuracy on our new RNA flow cells (e.g. FLO-MIN004RA). The flow cell priming reagents are also included as part of the sequencing kit and have been reformulated to be compatible on the new RNA flow cell. Other updates include fuel fix technology, allowing users to run longer experiments without the need for fuel addition during the run. Note: this kit is compatible with RNA-specific flow cells only (e.g. FLO-MIN004RA).
Input requirements: 200–300 ng poly(A)+ RNA or 1 µg total RNA.
In applications where direct RNA sequencing’s key advantages (detection of modified bases and reduced bias) are not important, users may consider one of Oxford Nanopore’s cDNA kits which currently provide the highest output per run.
This product includes a flow cell priming kit.
Delivery information: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.