Ice-Pak™ Foam Bricks, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
FBRK00002 FBRK00019 FBRK00001
76499-190CS 87.61 CAD
76499-190 76499-192 76499-194
Ice-Pak™ Foam Bricks, Cryopak Verification Technologies
Cooling Packs
Vacuum-sealed in durable film, these bricks are ideal for defined and precise packaging configurations.
  • Durable materials
  • Reusable
  • Consistent structure
Ice-Pak™ foam bricks have a freezing/melting point of 0 °C and offer superior thermal performance when transporting temperature-sensitive products. They feature a consistent, uniform structure that is maintained even after freezing.