TimeSaver™ Pre-Qualified Shippers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
FEPS01369 FEPS00820 FEPS00706 FEPS00630 FEPS00351
76499-166KT 75.51 CAD
76499-166 76499-168 76499-170 76499-172 76499-174
TimeSaver™ Pre-Qualified Shippers, Cryopak Verification Technologies
Mailers and Shippers
Pre-qualified shipping solutions are tested to the highest quality standards to preserve the integrity of transported products.
  • Packout instructions to ensure performance
  • Abridged qualification report
  • Qualification certificate
The TimeSaver™ solutions protect products that must be shipped within a specific temperature range for up to 24, 36, or 48 hours.