TemPak™ Insulated Shippers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
FEPS00040 FOM00033 FOM00024 FOM00031 FOM00023
76499-156EA 39.96 CAD
76499-156 76499-158 76499-160 76499-162 76499-164
TemPak™ Insulated Shippers, Cryopak Verification Technologies
Mailers and Shippers
Insulated shippers are the most cost-effective solution for same-day and next-day perishable shipments.
  • Durable outer carton
  • Custom printing available
Constructed with e×panded polystyrene (EPS) insulation panels, these packages can be used to ship a wide variety of applications within the cold chain.