CF® Dye Alkyne, Biotium
Supplier: Biotium
CF® Dye alkynes react with azide groups via copper-catalyzed bioorthogonal cycloaddition. Can be used to fluorescently detect or label azide groups on target molecules. CF® Dye alkynes can also be used as building blocks to form fluorescent polymers.
- Fluorescent detection or labeling azide groups on target molecules
- Can be used as fluorescent monomers to form dye-labeled polymers
- Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labeling
CF® Dye alkynes can react with azide groups to form 1,2,3-triazole via 1,3-dipolar Huisgen copper-catalyzed cycloaddition. They can also be used as monomeric building blocks to copolymerize with other monomers to form fluorescent polymers.
CF® Dyes are Biotium's line of next-generation fluorescent dyes that have improved brightness, photostability and water solubility compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.