Chromaster HPLC System, Hitachi
Supplier: Hitachi
Chromaster
892-0306 889-0136 892-0126 892-0787 892-0446 892-0606 892-0116 892-0226 892-0106 892-0406 892-0806 892-0797 892-0286 892-0486 892-0136 892-0426
76493-128EA 32604.49 CAD
76493-128 76493-148 76493-026 76493-146 76493-058 76493-178 76493-144 76493-132 76493-176 76493-142 76493-174 76493-064 76493-062 76493-060 76493-092 76493-180
Chromaster HPLC System, Hitachi
Chromatography Systems Complete HPLC Systems
The Chromaster HPLC system has established new limits in performance, ease of use, and robustness. Its design incorporates an extremely low dwell volume and a very low carry-over. In combination with very sensitive detector technology, especially the fluorescence detector, the Chromaster HPLC system is one of the strongest LC systems on the market. A huge amount of accessories and a modular set-up, enable an easy adaption to all customer needs, whether if it is used for an aggressive industrial or a high sensitive pharma application. VWR's relationship with Hitachi started over 30 years ago as a Merck Hitachi cooperation. During this time, the robustness and reliability of the Hitachi HPLC instruments has helped to build and develop a strong partnership.
  • Strong 8702.26 psi (60 MPa) pump
  • Excellent gradient reproducibility
  • Very low carry-over autosamplers
  • Loop injection and direct injection available with Peltier-thermostatted sample racks
  • Highly sensitive fluorescence detector (optimised 30 nm slit)
  • Very low drift diode array detector (1.0×10⁻⁴ AU hr⁻¹)
  • Automatic wavelength check using mercury lamp for UV, DAD and fluorescence detectors
  • Drivers available for OpenLab® CDS, Chromeleon® 6.8 and 7.X, Empower2® also Empower3® and Clarity
All pre-configured systems (with or without detector) are delivered with all necessary cables and an interface board.