Extra Large Cutters, 7260, Excelta
Supplier: Excelta
7260E 7261E
76471-622EA 174.97 CAD
76471-622 76471-624
Extra Large Cutters, 7260, Excelta
Cutters
The 7260 Series extra large oval cutter has a cutting capacity of .005" to .08" soft wire.
  • High grade carbon steel
  • Induction hardened edges
  • Can be re-sharpened
Order Now
SPECIFICATIONS
Environmentally Preferable