Miniature Wire Cutters, Excelta
Supplier: Excelta
9231E 9231SL 9232E 9232SL 9238E 9238SL
76471-632EA 168.78 CAD
76471-632 76471-634 76471-636 76471-638 76471-640 76471-642
Miniature Wire Cutters, Excelta
Cutters
These high-quality cutters are designed for cutting small soft wire up to 250,000 times without resharpening.
  • Three head shapes for various accessibility issues
  • Three types of cuts available
  • 'Set-Screw' stop prevents overtravel
  • Induction hardened blades for longevity
  • Ergonomic handles/dual leaf springs
These Lazer Line® side cutters are made of carbon tool steel with induction hardened cutting blades to RHC 63-65. The CNC machining allows the joint surfaces to be perfect to allow for long life. The dual leaf springs provide a smooth action. TheyMore Product Information