Secura® NTEP-Certified Analytical Balances, Sartorius
Supplier: Sartorius
SECURA224-1NUS SECURA124-1NUS SECURA224-1NUS SECURA124-1NUS
76488-194EA 8737.67 CAD
76488-194 76488-196 CA76488-194 CA76488-196
Secura® NTEP-Certified Analytical Balances, Sartorius
Balances and Scales
These Secura® balances give you the security and peace of mind of knowing that you have done everything right.
  • 100% NTEP-compliant weighing
  • Legal for Trade
  • Highly accurate weighing results and operating convenience
  • Protection systems for complete reliability and regulatory compliance
  • 100% traceable, clear documentation with sample and batch identifiers
  • Real-time guidance prompts for levelling, automatic internal adjustment
Besides providing highly accurate weighing results and operating convenience, the Secura® range also features built-in protection systems for complete reliability and regulatory compliance – the safe and secure way.