Scissors, Relieved Blade, Excelta Corp®, Excelta Corporation
Supplier: Excelta
351 352 353 354 355 356 357 358 359
76471-558EA 75.79 CAD
76471-558 76471-560 76471-562 76471-564 76471-566 76471-568 76471-570 76471-572 76471-574
Scissors, Relieved Blade, Excelta Corp®, Excelta Corporation
Scissors
These extremely precise relieved blade tempered stainless steel (AISI 420) scissors are ideal for delicate applications.
Order Now
SPECIFICATIONS
Environmentally Preferable