Disposable PE Transfer Pipettes, United Scientific Supplies
Supplier: United Scientific Supplies
F1016-5 F1016-9-PK/100 F1016-5 F1016-9-PK/100 F1016-7-PK/100
470332-796PK 45.15 CAD
470332-796 470332-798 470334-434 470334-436 470334-438
Disposable PE Transfer Pipettes, United Scientific Supplies
Pipettes Pasteur/Transfer Pipettes
All-in-one pipettes.
  • Molded
  • Unbreakable
  • Translucent
  • Pack of 100
All-in-one pipettes are molded from unbreakable translucent polyethylene. These versitle pipettes can be sealed and refrigerated. Perfect for classroom and laboratory use.