Gravity Convection Drying Ovens, Cole Parmer
Supplier: Antyila Scientific
52411-00 52411-02 52411-04 52411-06
76492-742EA 1386.24 CAD
76492-742 76492-744 76492-746 76492-748
Gravity Convection Drying Ovens, Cole Parmer
Ovens Standard Ovens
Low energy consumption provides operating cost savings.
  • Stainless steel inner chamber resists corrosion
  • Rounded inner chamber corners offer easy cleaning
  • Samples can be heated up to 392 °F (200 °C)
Gravity convection ovens feature a PID controller which provides accurate and reliable temperature control up to 392 °F (200 °C). Natural convection heating allows sample temperature to be uniform; suitable for sterilization, powder drying, and high-temperature storage. The ovens require minimal heating time, reaching 212 °F (100 °C) in 15 to 30 minutes.More Product Information