Deuterium Lamp, Gilson/Rainin 115, 116, 117, Optimize Technologies
Supplier: Optimize Technologies
26-32-03671
89522-424EA 4122.99 CAD
89522-424
Deuterium Lamp, Gilson/Rainin 115, 116, 117, Optimize Technologies
Lights Deuterium Lamps
Optimize Technologies offers a broad selection of lamps for OEM HPLC detectors.
  • 1000 Hour lifetime guarantee on all Standard Lamps
  • 2000 Hour lifetime guarantee on all DX2 DoubleLife
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee.
Ordering information: Need a lamp that is not listed on our website? Please contact us.