Dual Conductor Speidel Metal Wrist Strap, Transforming Technologies
Supplier: Transforming Technologies
WB0050L WB0050M WB0050S
76490-228EA 48.05 CAD
76490-228 76490-230 76490-232
Dual Conductor Speidel Metal Wrist Strap, Transforming Technologies
Grounders Wrist Grounders
The Speidel metal band features two 180 degree conductive paths around the wrist, providing maximum skin contact.
  • Premium metal expansion wrist strap with more reliable path-to-ground
  • Durable metal band is designed for long wearing, and is easy to clean
  • Dual-Wire conductor provides redundancy
  • If one dual-wire wrist strap wire is severed, operator has reliable path-to-ground with other wire
The expandable band is constructed with two stainless steel back plates and is available in small, medium, and large sizes.