BFN AC Ionizer Spare Parts, Transforming Technologies
Supplier: Transforming Technologies
SPBFN10 SPBFN11 SPBFN13 SPBFN14 SPBFN15 SPBFN16 SPBFN17 SPBFN18 SPBFN19 SPBFN20 SPBFN21 SPBFN22 SPBFN23 SPBFN24 SPBFN26 SPBFN28 SPBFN29 SPBFN35
76490-180EA 51.82 CAD
76490-180 76490-182 76490-184 76490-186 76490-188 76490-190 76490-192 76490-194 76490-196 76490-198 76490-200 76490-202 76490-204 76490-206 76490-208 76490-210 76490-212 76490-214
BFN AC Ionizer Spare Parts, Transforming Technologies
Ionizers
Use the following spare parts for BFN Ionizer models: BFN801, BFN802, BFN803, and BFN8412.
  • Replacement parts for BFN series Ionizers