Air Track Gliders
Supplier: Wards
PH0362GLD2
470224-382EA 225.45 CAD
470224-382
Air Track Gliders
Educational Materials Physics Educational Materials Mechanics Learning Activities
A pair of 100g gliders designed for use with air tracks 470157-654 and 470157-664.
  • Made from anodized aluminum
  • Each glider has flanks that are perpendicular to each other and sit 45 degrees below the horizontal
  • Trails along the side of the glider allow for the attachment of masses, flags, or other objects
  • Gliders can be used to simulate motion and kinematics in a near-frictionless environment on the air track
High quality 100g gliders designed with unique features including the ability to add masses, flags, or other objects. Compatible with a varietyMore Product Information