Eisco Premium Glass Double Concave Lenses
Supplier: Wards
PH0537GCC PH0537HCC PH0537ICC PH0537JCC PH0537KCC
470328-114EA 30.95 CAD
470328-114 470328-116 470328-118 470328-120 470328-122
Eisco Premium Glass Double Concave Lenses
Educational Materials Physics Educational Materials Optics Learning Activities
Premium line of glass lenses designed specifically for Universities.
  • Made of high quality, extra white glass
  • Highly polished, double concave glass lenses
  • Highly polished with ground edges
  • Spherical
Premium line of glass lenses designed for advanced studies and demonstrations in optics and light refraction. Because of their educational use design, there may be minor imperfections that do not affect physical function.
Order Now
SPECIFICATIONS
Environmentally Preferable