Glass Concave Mirror
Supplier: Wards
PH0519A PH0519C PH0519D PH0520A PH0520B PH0520H PH0520I PH0520J PH0520K PH0520L PH0520M PH0520N PH0520O PH0520P
470328-816EA 4.4 CAD
470328-816 470328-818 470328-820 470328-822 470328-824 470328-826 470328-828 470328-830 470328-832 470328-834 470328-836 470328-838 470328-840 470328-842
Glass Concave Mirror
Mirrors
Round concave glass mirror designed for physics and educational uses.
  • Round, high-quality glass mirror
  • Optically worked
  • Silvered back with lead -free protective coating
Designed for physics and educational applications. Because of their educational use design, there may be minor imperfections that do not affect physical function.