Universal Reverse Primer, Agilent Technologies

Supplier: Agilent Technologies
600037 600037
CA97066-434EA 199.82 CAD
CA97066-434 97066-434
Universal Reverse Primer, Agilent Technologies
Nucleic Acid Reagents Primers and Probes
The universal reverse primer serves as the downstream primer in the qPCR reaction by annealing to a universal sequence tag that is added to the cDNA during the reverse transcription protocol.

Applications: PCR
Number of Reactions: 3.125 μM
Physical Form: Liquid
Reactions per Kit: 300
Volume: 200 µL
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR