Extracta Plus DNA Kits, Quantabio
Supplier: Quantabio
95213-010 95213-050
CA76492-578EA 61.67 CAD
CA76492-578 CA76492-580
Extracta Plus DNA Kits, Quantabio
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Rapid extraction and purification of high-quality total DNA from cultured cells or tissue.
  • High yield – Obtain large quantities of high quality DNA, free from PCR inhibitors
  • Versatility in sample types – Rapid purification of fresh or frozen tissue, cells, blood or bacteria
  • Lab friendly – Silica based column eliminates toxic organic chemicals such as phenol/chloroform
  • Downstream compatibility – Ideal for sensitive applications such as endpoint PCR, qPCR and next generation sequencing
Extracta Plus DNA kits offer rapid purification of total DNA from a variety of sample sources. Genomic, mitochondrial and pathogenic DNA can be collected from fresh/frozen animal cells, tissue, blood, and bacteria. The Extracta Plus Spin Column workflow enables simultaneous processing of multiple samples into purified DNA suitable for PCR, Southern blotting, RAPD, AFLP and next generation sequencing applications.
High yielding gDNA extraction from multiple sources Extracta Plus DNA allows high yielding extraction of total DNA (example: genomic, mitochondrial and pathogen) from various tissues as well as cultured cells and blood samples.
Extracted DNA is free of inhibitors that may impact downstream applications such as PCR, qPCR and next generation sequencing.
It allows high molecular weight gDNA extraction, enabling long range PCR and sequencing applications.