Sodium hexametaphosphate, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium metaphosphateSHMP

BAKRV025-QM BAKRV025-05 BAKRV025-05 BAKRV025-QM
CAJTV025-QMEA 249.78 CAD
CAJTV025-QM JTV025-QM CAJTV025-05 JTV025-05
Sodium hexametaphosphate, Biotech Reagent, J.T.Baker®
Sodium hexametaphosphate
Formula: Na₆P₆O₁₈
MW: 611.77 g/mol
MDL Number: MFCD00136045
CAS Number: 10124-56-8

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Identification Passes Test
Appearance (white to off-white solid) Passes Test
pH of 1% Solution at 25°C 6.7 - 7.2
Loss on Drying at 105°C ≤ 0.5 %
Assay (as P₂O₅) 60.0 - 71.0 %
Endotoxin Concentration (EU/g), For Information Only
Total Aerobic Microbial Count ≤ 100 cfu/g
Total Yeasts and Molds ≤ 100 cfu/g
Residual Methanol ≤ 3000 ppm
Residual Ethanol ≤ 5000 ppm
Residual Isopropanol ≤ 5000 ppm
Aluminum (Al), For Information Only
Antimony (Sb), For Information Only
Arsenic (As), For Information Only
Barium (Ba), For Information Only
Cadmium (Cd), For Information Only
Chromium (Cr), For Information Only
Cobalt (Co), For Information Only
Copper (Cu), For Information Only
Gold (Au), For Information Only
Iridium (Ir), For Information Only
Lithium (Li), For Information Only
Manganese (Mn), For Information Only
Mercury (Hg), For Information Only
Molybdenum (Mo), For Information Only
Nickel (Ni), For Information Only
Osmium (Os), For Information Only
Palladium (Pd), For Information Only
Platinum (Pt), For Information Only
Rhodium (Rh), For Information Only
Ruthenium (Ru), For Information Only
Selenium (Se), For Information Only
Silver (Ag), For Information Only
Thallium (Tl), For Information Only
Tin (Sn), For Information Only
Vanadium (V), For Information Only
Zinc (Zn), For Information Only
Residual solvents - Only the Class 2 solvent methanol and Class 3
solvents ethanol and isopropanol are likely to be present as a result of
the production process. The residual concentrations are determined and
reported for each production batch.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR